The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167890.1 | NP_001161362.1 | 1 - 2 | 365 | 60 |
NM_015507.3 | NP_056322.2 | 1 - 2 | 365 | 60 | |
GenBank mRNA | AF186084.1 | - | 1 - 2 | 223 | 60 |
AF193055.1 | - | 1 - 2 | 291 | 60 | |
AJ245671.1 | - | 1 - 2 | 318 | 60 | |
AK075214.1 | - | 1 - 2 | 335 | 60 | |
AK300236.1 | - | 1 - 2 | 335 | 60 | |
AK315018.1 | - | 1 - 2 | 203 | 60 | |
AY358333.1 | - | 1 - 2 | 212 | 60 | |
BC038587.1 | - | 1 - 2 | 174 | 60 | |
DA534733.1 | - | 1 - 2 | 336 | 60 | |
DC380958.1 | - | 1 - 2 | 365 | 60 |