The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006682.2 | NP_006673.1 | 1 - 2 | 651 | 88 |
GenBank mRNA | AK222999.1 | - | 1 - 2 | 619 | 88 |
AK312672.1 | - | 1 - 2 | 623 | 88 | |
BC033820.1 | - | 1 - 2 | 639 | 88 | |
BC073986.1 | - | 1 - 2 | 624 | 88 | |
Z36531.1 | - | 1 - 2 | 644 | 88 |