The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001150.2 | NP_001141.2 | 20 - 21 | 3043 | 64 |
XM_005254892.4 | XP_005254949.1 | 20 - 21 | 3051 | 64 | |
XM_011521473.1 | XP_011519775.1 | 20 - 21 | 3194 | 64 | |
GenBank mRNA | AB209918.1 | - | 20 - 21 | 3039 | 64 |
AK298134.1 | - | 14 - 15 | 1513 | 64 | |
AK298242.1 | - | 14 - 15 | 1564 | 64 | |
AK298339.1 | - | 15 - 16 | 1624 | 64 | |
AK300946.1 | - | 16 - 17 | 1774 | 64 | |
BC058928.1 | - | 19 - 20 | 2844 | 64 | |
M22324.1 | - | 19 - 20 | 2871 | 64 | |
X13276.1 | - | 19 - 20 | 2775 | 64 |