The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126054.2 | NP_001119526.1 | 20 - 21 | 2020 | 100 |
NM_001126055.2 | NP_001119527.1 | 19 - 20 | 2002 | 100 | |
NM_003688.3 | NP_003679.2 | 21 - 22 | 2089 | 100 | |
XM_005272686.4 | XP_005272743.1 | 20 - 21 | 2562 | 100 | |
XM_006724566.3 | XP_006724629.1 | 18 - 19 | 2457 | 100 | |
XM_011543993.2 | XP_011542295.1 | 20 - 21 | 2141 | 100 | |
XM_011543994.2 | XP_011542296.1 | 19 - 20 | 2105 | 100 | |
XM_011543995.2 | XP_011542297.1 | 19 - 20 | 2072 | 100 | |
XM_011543996.2 | XP_011542298.1 | 18 - 19 | 2036 | 100 | |
XM_011543997.2 | XP_011542299.1 | 15 - 16 | 1800 | 100 | |
GenBank mRNA | AB039327.2 | - | 8 - 9 | 1178 | 100 |
AB208859.1 | - | 21 - 22 | 2033 | 100 | |
AF032119.1 | - | 21 - 22 | 2070 | 100 | |
AF035582.1 | - | 19 - 20 | 1971 | 100 | |
AF262404.1 | - | 16 - 17 | 1529 | 100 | |
AF262405.1 | - | 18 - 19 | 1878 | 100 | |
AK313399.1 | - | 19 - 20 | 2155 | 100 | |
AY705392.1 | - | 21 - 22 | 2049 | 100 | |
BC117311.1 | - | 20 - 21 | 1993 | 100 | |
BC143454.1 | - | 20 - 21 | 1993 | 100 | |
BC143456.1 | - | 19 - 20 | 1975 | 100 |