Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRK4
Entrez Gene ID
2868
Gene Name
G protein-coupled receptor kinase 4
Gene Aliases
GPRK2L, GPRK4, GRK4a, IT11
Chromosome Location
Chr.4: 2963504 - 3040760 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004056.1 NP_001004056.1 9 - 10 1328 104
NM_001004057.1 NP_001004057.1 10 - 11 1424 104
NM_005307.2 NP_005298.2 9 - 10 1328 104
NM_182982.2 NP_892027.2 10 - 11 1424 104
XM_005247962.3 XP_005248019.1 8 - 9 942 104
XM_006713880.3 XP_006713943.1 9 - 10 840 104
XM_011513447.2 XP_011511749.1 11 - 12 1653 104
XM_011513448.2 XP_011511750.1 11 - 12 1653 104
XM_011513449.2 XP_011511751.1 10 - 11 1557 104
XM_011513450.2 XP_011511752.1 11 - 12 1653 104
XM_011513451.2 XP_011511753.1 11 - 12 1653 104
XM_011513452.2 XP_011511754.1 11 - 12 1653 104
XM_011513453.2 XP_011511755.1 11 - 12 1653 104
XM_011513454.2 XP_011511756.1 11 - 12 1653 104
XM_011513455.2 XP_011511757.1 11 - 12 1653 104
XM_017008052.1 XP_016863541.1 10 - 11 1575 104
XM_017008054.1 XP_016863543.1 10 - 11 1536 104
XM_017008055.1 XP_016863544.1 9 - 10 1440 104
XM_017008056.1 XP_016863545.1 10 - 11 1536 104
XM_017008057.1 XP_016863546.1 7 - 8 740 104
XM_017008058.1 XP_016863547.1 9 - 10 800 104
XM_017008059.1 XP_016863548.1 7 - 8 824 104
XM_017008060.1 XP_016863549.1 8 - 9 918 104
XM_017008061.1 XP_016863550.1 9 - 10 1191 104
XM_017008062.1 XP_016863551.1 9 - 10 1338 104
XM_017008063.1 XP_016863552.1 8 - 9 1127 104
XM_017008064.1 XP_016863553.1 8 - 9 911 104
XM_017008066.1 XP_016863555.1 7 - 8 748 104
GenBank mRNA AK223581.1 - 9 - 10 1336 104
AK292390.1 - 10 - 11 1469 104
BC117320.1 - 10 - 11 1312 104
U33054.1 - 10 - 11 1223 104
U33055.1 - 9 - 10 1127 104
U33056.1 - 10 - 11 1223 104
X97879.1 - 9 - 10 971 104
X97880.1 - 9 - 10 971 104
X97881.1 - 10 - 11 1067 104
X98118.1 - 10 - 11 1067 104

View Full Product Details