The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004815.3 | NP_004806.3 | 22 - 23 | 3275 | 69 |
XM_006711048.3 | XP_006711111.1 | 21 - 22 | 2909 | 69 | |
XM_011542438.2 | XP_011540740.1 | 22 - 23 | 3150 | 69 | |
XM_011542439.2 | XP_011540741.1 | 22 - 23 | 2995 | 69 | |
XM_011542440.2 | XP_011540742.1 | 21 - 22 | 2869 | 69 | |
GenBank mRNA | AB208873.1 | - | 21 - 22 | 2881 | 69 |
AK310491.1 | - | 22 - 23 | 2995 | 69 | |
BC093741.1 | - | 21 - 22 | 2948 | 69 | |
BC093767.1 | - | 21 - 22 | 2951 | 69 | |
CB852768.1 | - | 3 - 4 | 483 | 69 | |
U90920.1 | - | 22 - 23 | 3089 | 69 |