The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005244.4 | NP_005235.3 | 2 - 3 | 481 | 79 |
NM_172110.3 | NP_742108.2 | 2 - 3 | 481 | 79 | |
XM_005260327.2 | XP_005260384.1 | 2 - 3 | 174 | 79 | |
XM_017027721.1 | XP_016883210.1 | 2 - 3 | 199 | 79 | |
GenBank mRNA | AK293032.1 | - | 2 - 3 | 185 | 79 |
AY705349.1 | - | 1 - 2 | 117 | 79 | |
BC008803.2 | - | 2 - 3 | 235 | 79 | |
BC013882.2 | - | 2 - 3 | 481 | 79 | |
BT006682.1 | - | 1 - 2 | 107 | 79 | |
U71207.1 | - | 1 - 2 | 109 | 79 |