The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005829.4 | NP_005820.1 | 1 - 2 | 505 | 73 |
NR_023361.1 | 1 - 2 | 505 | 73 | ||
GenBank mRNA | AK222698.1 | - | 1 - 2 | 97 | 73 |
AK294712.1 | - | 1 - 2 | 97 | 73 | |
AK312467.1 | - | 1 - 2 | 129 | 73 | |
BC002785.2 | - | 1 - 2 | 90 | 73 | |
BC010020.2 | - | 1 - 2 | 117 | 73 | |
DC340463.1 | - | 1 - 2 | 114 | 73 | |
DC410373.1 | - | 1 - 2 | 506 | 73 | |
X99459.1 | - | 1 - 2 | 99 | 73 |