Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PROP1
Entrez Gene ID
5626
Gene Name
PROP paired-like homeobox 1
Gene Aliases
CPHD2, PROP-1
Chromosome Location
Chr.5: 177992235 - 177996242 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006261.4 NP_006252.3 1 - 2 415 94
GenBank mRNA AF076215.1 - 1 - 2 415 94
BC069076.1 - 1 - 2 106 94

View Full Product Details