The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293306.2 | NP_001280235.2 | 11 - 12 | 1755 | 107 |
NM_006514.3 | NP_006505.3 | 11 - 12 | 1755 | 107 | |
XM_005265371.3 | XP_005265428.1 | 11 - 12 | 1764 | 107 | |
XM_011533993.2 | XP_011532295.1 | 11 - 12 | 1764 | 107 | |
GenBank mRNA | AF117907.1 | - | 11 - 12 | 1755 | 107 |