The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006862.3 | NP_006853.2 | 13 - 14 | 1910 | 62 |
XM_017000126.1 | XP_016855615.1 | 13 - 14 | 1851 | 62 | |
XM_017000127.1 | XP_016855616.1 | 13 - 14 | 1880 | 62 | |
XM_017000130.1 | XP_016855619.1 | 13 - 14 | 1744 | 62 | |
GenBank mRNA | AF119121.2 | - | 13 - 14 | 1898 | 62 |
BC032690.1 | - | 13 - 14 | 1817 | 62 |