The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012384.4 | NP_036516.1 | 6 - 7 | 824 | 91 |
XM_005260202.4 | XP_005260259.1 | 6 - 7 | 1035 | 91 | |
XM_006723776.2 | XP_006723839.1 | 4 - 5 | 476 | 91 | |
XM_011528779.2 | XP_011527081.1 | 4 - 5 | 540 | 91 | |
GenBank mRNA | AB033095.1 | - | 6 - 7 | 701 | 91 |
AF173867.1 | - | 5 - 6 | 1100 | 91 | |
AL133646.1 | - | 4 - 5 | 548 | 91 | |
BC036305.1 | - | 6 - 7 | 751 | 91 | |
BQ954281.1 | - | 5 - 6 | 526 | 91 |