The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001280796.1 | NP_001267725.1 | 1 - 2 | 476 | 73 |
NM_001280797.1 | NP_001267726.1 | 1 - 2 | 476 | 73 | |
NM_013267.3 | NP_037399.2 | 1 - 2 | 476 | 73 | |
GenBank mRNA | AF110330.1 | - | 1 - 2 | 231 | 73 |
AF223944.1 | - | 1 - 2 | 403 | 73 | |
AK000467.1 | - | 1 - 2 | 223 | 73 | |
AK289471.1 | - | 1 - 2 | 180 | 73 | |
AK294822.1 | - | 1 - 2 | 421 | 73 | |
AK303772.1 | - | 1 - 2 | 311 | 73 | |
BC025661.1 | - | 1 - 2 | 191 | 73 | |
BC119767.1 | - | 1 - 2 | 421 | 73 | |
BC119768.1 | - | 1 - 2 | 421 | 73 |