The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013283.4 | NP_037415.1 | 5 - 6 | 878 | 68 |
NM_182796.2 | NP_877725.1 | 5 - 6 | 1017 | 68 | |
GenBank mRNA | AF113225.1 | - | 5 - 6 | 674 | 68 |
AF182814.1 | - | 5 - 6 | 719 | 68 | |
AJ243721.1 | - | 5 - 6 | 857 | 68 | |
AK096178.1 | - | 4 - 5 | 2138 | 68 | |
AK291919.1 | - | 5 - 6 | 828 | 68 | |
AK294069.1 | - | 4 - 5 | 532 | 68 | |
AK312365.1 | - | 5 - 6 | 827 | 68 | |
AL136664.1 | - | 5 - 6 | 791 | 68 | |
AY358695.1 | - | 5 - 6 | 772 | 68 | |
BC005218.1 | - | 5 - 6 | 798 | 68 | |
BC066645.1 | - | 5 - 6 | 753 | 68 | |
BC093030.1 | - | 5 - 6 | 787 | 68 | |
DQ413183.1 | - | 5 - 6 | 856 | 68 |