The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013391.3 | NP_037523.2 | 14 - 15 | 2303 | 65 |
GenBank mRNA | AF111858.1 | - | 14 - 15 | 2249 | 65 |
AK057641.1 | - | 14 - 15 | 2255 | 65 | |
AK314736.1 | - | 14 - 15 | 2261 | 65 |