The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014275.4 | NP_055090.1 | 1 - 2 | 467 | 87 |
XM_017008998.1 | XP_016864487.1 | 1 - 2 | 293 | 87 | |
GenBank mRNA | AB000624.1 | - | 1 - 2 | 140 | 87 |
AY358984.1 | - | 1 - 2 | 148 | 87 | |
BC051835.1 | - | 1 - 2 | 343 | 87 |