The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317381.1 | NP_001304310.1 | 4 - 5 | 641 | 67 |
NM_001317382.1 | NP_001304311.1 | 3 - 4 | 360 | 67 | |
NM_014278.3 | NP_055093.2 | 3 - 4 | 651 | 67 | |
XM_011531745.2 | XP_011530047.1 | 3 - 4 | 720 | 67 | |
GenBank mRNA | AB023421.1 | - | 3 - 4 | 363 | 67 |
AK302115.1 | - | 3 - 4 | 334 | 67 | |
AK303053.1 | - | 4 - 5 | 641 | 67 | |
AY290862.1 | - | 3 - 4 | 517 | 67 | |
BC040560.1 | - | 3 - 4 | 562 | 67 | |
DB086945.1 | - | 3 - 4 | 360 | 67 | |
EF197155.1 | - | 3 - 4 | 309 | 67 |