Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN4Y
Entrez Gene ID
22829
Gene Name
neuroligin 4, Y-linked
Gene Aliases
-
Chromosome Location
Chr.Y: 14522578 - 14844945 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206850.1 NP_001193779.1 1 - 2 333 65
GenBank mRNA AB023168.1 - 1 - 2 189 65

View Full Product Details