The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_197962.2 | NP_932066.1 | 1 - 2 | 215 | 101 |
GenBank mRNA | AF132495.2 | - | 1 - 2 | 130 | 101 |
AF151891.1 | - | 1 - 2 | 105 | 98 | |
AY038988.1 | - | 1 - 2 | 137 | 101 | |
BC009669.1 | - | 1 - 2 | 130 | 101 | |
BC028113.1 | - | 1 - 2 | 121 | 101 | |
BU157087.1 | - | - | 170 | 101 | |
CN362908.1 | - | 1 - 2 | 239 | 101 |