The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016204.2 | NP_057288.1 | 1 - 2 | 526 | 62 |
GenBank mRNA | AF188285.1 | - | 1 - 2 | 510 | 62 |
AK314956.1 | - | 1 - 2 | 526 | 62 | |
BC069643.1 | - | 1 - 2 | 429 | 62 | |
BC074921.2 | - | 1 - 2 | 378 | 62 |