The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185054.1 | NP_001171983.1 | 2 - 3 | 228 | 72 |
NM_001617.3 | NP_001608.1 | 2 - 3 | 474 | 72 | |
NM_017482.3 | NP_059516.2 | 2 - 3 | 474 | 72 | |
NM_017488.3 | NP_059522.1 | 2 - 3 | 474 | 72 | |
XM_011532502.2 | XP_011530804.1 | 2 - 3 | 215 | 72 | |
GenBank mRNA | AB209227.1 | - | 2 - 3 | 443 | 72 |
AK291007.1 | - | 2 - 3 | 429 | 72 | |
BC010237.1 | - | 2 - 3 | 408 | 72 | |
BC041666.1 | - | 2 - 3 | 408 | 72 | |
BC051882.1 | - | 2 - 3 | 401 | 72 | |
BC056881.1 | - | 2 - 3 | 257 | 72 | |
BC065525.1 | - | 2 - 3 | 228 | 72 | |
CN428088.1 | - | 2 - 3 | 429 | 72 |