The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017688.2 | NP_060158.2 | 3 - 4 | 583 | 66 |
GenBank mRNA | AJ276691.1 | - | 3 - 4 | 564 | 66 |
AK000157.1 | - | 3 - 4 | 309 | 66 | |
AK092607.1 | - | 3 - 4 | 574 | 66 | |
BX462607.2 | - | 3 - 4 | 582 | 66 |