Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN3
Entrez Gene ID
54413
Gene Name
neuroligin 3
Gene Aliases
HNL3
Chromosome Location
Chr.X: 71144389 - 71175307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166660.1 NP_001160132.1 1 - 2 133 101
NM_018977.3 NP_061850.2 1 - 2 133 101
NM_181303.1 NP_851820.1 1 - 2 133 101
XM_006724663.3 XP_006724726.1 1 - 2 575 101
GenBank mRNA AF217411.1 - 1 - 2 94 101
BC028738.1 - 1 - 2 128 101
BC051715.1 - 1 - 2 63 101
BP201014.1 - 1 - 2 133 101

View Full Product Details