The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020169.3 | NP_064554.3 | 1 - 2 | 337 | 135 |
XM_017006876.1 | XP_016862365.1 | 1 - 2 | 366 | 135 | |
GenBank mRNA | AF087851.1 | - | 1 - 2 | 200 | 135 |
AF282594.1 | - | 1 - 2 | 206 | 135 | |
AF282626.1 | - | 1 - 2 | 273 | 135 | |
BC005346.1 | - | 1 - 2 | 337 | 135 | |
BC008438.1 | - | 1 - 2 | 319 | 135 |