The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021080.3 | NP_066566.3 | 6 - 7 | 984 | 140 |
GenBank mRNA | AB210012.1 | - | 6 - 7 | 966 | 140 |
AK095513.1 | - | 6 - 7 | 984 | 140 | |
AK313144.1 | - | 4 - 5 | 631 | 140 | |
BC067445.1 | - | 3 - 4 | 346 | 140 | |
BC067446.1 | - | 3 - 4 | 346 | 140 | |
BC112938.1 | - | 4 - 5 | 953 | 140 |