The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024647.3 | NP_001019818.1 | 4 - 5 | 612 | 89 |
NM_001278402.1 | NP_001265331.1 | 4 - 5 | 527 | 89 | |
NM_022456.4 | NP_071901.2 | 6 - 7 | 1263 | 89 | |
NM_175623.3 | NP_783322.1 | 6 - 7 | 1397 | 89 | |
NM_175624.3 | NP_783323.1 | 6 - 7 | 1263 | 89 | |
NM_175625.3 | NP_783324.1 | 6 - 7 | 1397 | 89 | |
XM_006719226.3 | XP_006719289.3 | 6 - 7 | 1386 | 89 | |
XM_006719227.3 | XP_006719290.3 | 6 - 7 | 1059 | 89 | |
XM_017018773.1 | XP_016874262.1 | 6 - 7 | 1021 | 89 | |
XM_017018774.1 | XP_016874263.1 | 8 - 9 | 1274 | 89 | |
XM_017018776.1 | XP_016874265.1 | 7 - 8 | 1146 | 89 | |
XM_017018777.1 | XP_016874266.1 | 7 - 8 | 1565 | 89 | |
GenBank mRNA | AJ312896.1 | - | 6 - 7 | 1087 | 89 |
AJ312897.1 | - | 6 - 7 | 1379 | 89 | |
AJ312898.1 | - | 6 - 7 | 1087 | 89 | |
AJ312899.1 | - | 6 - 7 | 1379 | 89 | |
AK026201.1 | - | 6 - 7 | 1087 | 89 | |
AK027566.1 | - | 4 - 5 | 523 | 89 | |
AK027671.1 | - | 4 - 5 | 513 | 89 | |
AK057844.1 | - | 6 - 7 | 992 | 89 | |
AK225441.1 | - | 6 - 7 | 1087 | 89 | |
AK314547.1 | - | 6 - 7 | 1098 | 89 | |
BC015548.1 | - | 4 - 5 | 590 | 89 | |
DN912041.1 | - | 4 - 5 | 612 | 89 |