The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022744.3 | NP_073581.2 | 6 - 7 | 751 | 64 |
XM_017023570.1 | XP_016879059.1 | 4 - 5 | 582 | 64 | |
GenBank mRNA | AK000679.1 | - | 2 - 3 | 172 | 64 |
AK023930.1 | - | 6 - 7 | 717 | 64 | |
AK222913.1 | - | 6 - 7 | 717 | 64 | |
AK296166.1 | - | 5 - 6 | 601 | 64 | |
AK296491.1 | - | 4 - 5 | 579 | 64 | |
AK304608.1 | - | 7 - 8 | 579 | 64 | |
BC009308.2 | - | 6 - 7 | 711 | 64 |