The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320147.1 | NP_001307076.1 | 1 - 2 | 463 | 71 |
NM_024046.4 | NP_076951.2 | 1 - 2 | 463 | 71 | |
GenBank mRNA | AK074856.1 | - | 1 - 2 | 177 | 71 |
AK075294.1 | - | 1 - 2 | 158 | 71 | |
AK295588.1 | - | 1 - 2 | 150 | 71 | |
AK297258.1 | - | 1 - 2 | 147 | 71 | |
AK297437.1 | - | 1 - 2 | 144 | 71 | |
AK299953.1 | - | 1 - 2 | 146 | 71 | |
AK315525.1 | - | 1 - 2 | 196 | 71 | |
AL136697.1 | - | 1 - 2 | 146 | 71 | |
AL833943.1 | - | 1 - 2 | 147 | 71 | |
BC000497.2 | - | 1 - 2 | 132 | 71 | |
BC001921.1 | - | 1 - 2 | 132 | 71 | |
BC005828.2 | - | 1 - 2 | 132 | 71 | |
BC017363.2 | - | 1 - 2 | 134 | 71 | |
BC019256.1 | - | 1 - 2 | 127 | 71 | |
BX647074.1 | - | 1 - 2 | 144 | 71 | |
DC351900.1 | - | 1 - 2 | 463 | 71 |