The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001928.2 | NP_001001928.1 | 3 - 4 | 393 | 147 |
NM_005036.4 | NP_005027.2 | 4 - 5 | 477 | 147 | |
XM_005261655.3 | XP_005261712.1 | 4 - 5 | 770 | 147 | |
XM_005261656.3 | XP_005261713.1 | 5 - 6 | 934 | 147 | |
XM_006724269.3 | XP_006724332.1 | 4 - 5 | 536 | 147 | |
XM_006724270.3 | XP_006724333.1 | 3 - 4 | 442 | 147 | |
XM_011530239.2 | XP_011528541.1 | 5 - 6 | 626 | 147 | |
XM_011530240.2 | XP_011528542.1 | 5 - 6 | 629 | 147 | |
XM_011530241.2 | XP_011528543.1 | 4 - 5 | 534 | 147 | |
XM_011530242.2 | XP_011528544.1 | 3 - 4 | 356 | 147 | |
XM_011530243.2 | XP_011528545.1 | 2 - 3 | 597 | 147 | |
XM_011530244.2 | XP_011528546.1 | 5 - 6 | 629 | 147 | |
XM_011530245.2 | XP_011528547.1 | 3 - 4 | 445 | 147 | |
XM_017028840.1 | XP_016884329.1 | 5 - 6 | 629 | 147 | |
GenBank mRNA | AB307690.1 | - | 1 - 2 | 211 | 147 |
AK289821.1 | - | 4 - 5 | 770 | 147 | |
BC000052.2 | - | 3 - 4 | 421 | 147 | |
CR456547.1 | - | 2 - 3 | 347 | 147 | |
CR457435.1 | - | 1 - 2 | 211 | 147 | |
EU395809.1 | - | 1 - 2 | 211 | 147 | |
EU650667.1 | - | 1 - 2 | 211 | 147 | |
HQ692862.1 | - | 1 - 2 | 211 | 147 | |
L02932.1 | - | 3 - 4 | 427 | 147 | |
S74349.1 | - | 2 - 3 | 334 | 147 |