The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014553.2 | NP_055368.1 | 13 - 14 | 1435 | 68 |
XM_017003903.1 | XP_016859392.1 | 8 - 9 | 742 | 68 | |
GenBank mRNA | AF198488.1 | - | 13 - 14 | 1432 | 68 |
BC064698.1 | - | 13 - 14 | 1435 | 68 |