The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002983.2 | NP_002974.1 | 1 - 2 | 175 | 53 |
GenBank mRNA | BC071834.1 | - | 1 - 2 | 147 | 53 |
BC171831.1 | - | 1 - 2 | 109 | 53 | |
BC171891.1 | - | 1 - 2 | 109 | 53 | |
BX103455.1 | - | 1 - 2 | 175 | 53 | |
D00044.1 | - | 1 - 2 | 156 | 53 | |
D63785.1 | - | 1 - 2 | 56 | 53 | |
M23452.1 | - | 1 - 2 | 156 | 53 | |
M25315.1 | - | 1 - 2 | 156 | 53 | |
X03754.1 | - | 1 - 2 | 156 | 53 |