The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018556.3 | NP_061026.2 | 4 - 5 | 1148 | 65 |
XM_005260749.3 | XP_005260806.1 | 3 - 4 | 866 | 65 | |
XM_011529286.2 | XP_011527588.1 | 4 - 5 | 1195 | 65 | |
GenBank mRNA | AB042624.1 | - | 4 - 5 | 65 | |
AK292745.1 | - | 4 - 5 | 65 | ||
BC064532.1 | - | 4 - 5 | 65 |