The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019120.4 | NP_061993.3 | 1 - 1 | 705 | 144 |
GenBank mRNA | AF152501.2 | - | 1 - 1 | 460 | 144 |
BC136801.1 | - | 1 - 1 | 705 | 144 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018933.3 | NP_061756.1 | 1 - 1 | 144 | |
GenBank mRNA | AF152492.1 | - | 1 - 1 | 144 | |
AF217745.1 | - | 1 - 1 | 144 | ||
AK021915.1 | - | 1 - 1 | 144 | ||
AK292821.1 | - | 1 - 1 | 144 | ||
AK296151.1 | - | 1 - 1 | 144 | ||
AY358427.1 | - | 1 - 1 | 144 | ||
BC033068.1 | - | 1 - 1 | 144 | ||
BC051348.1 | - | 1 - 1 | 144 | ||
BC071934.1 | - | 1 - 1 | 144 |