The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319074.1 | NP_001306003.1 | 2 - 3 | 544 | 109 |
NM_032753.3 | NP_116142.1 | 2 - 3 | 285 | 109 | |
GenBank mRNA | AY211277.1 | - | 2 - 3 | 276 | 109 |
BC007284.1 | - | 2 - 3 | 542 | 109 | |
BC018709.1 | - | 2 - 3 | 544 | 109 | |
BC032512.1 | - | 2 - 3 | 555 | 109 |