The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316978.1 | NP_001303907.1 | 2 - 3 | 1824 | 110 |
NM_001316979.1 | NP_001303908.1 | 2 - 3 | 1551 | 110 | |
NM_001316980.1 | NP_001303909.1 | 2 - 3 | 1374 | 110 | |
NM_001316981.1 | NP_001303910.1 | 2 - 3 | 1676 | 110 | |
NM_001316982.1 | NP_001303911.1 | 2 - 3 | 1494 | 110 | |
NM_032792.3 | NP_116181.1 | 2 - 3 | 1580 | 110 | |
XM_006723445.3 | XP_006723508.1 | 2 - 3 | 1594 | 110 | |
GenBank mRNA | AK027392.1 | - | 2 - 3 | 1363 | 110 |
AL831909.1 | - | 1 - 2 | 125 | 110 | |
BC024738.1 | - | 2 - 3 | 1575 | 110 |