Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM222A
Entrez Gene ID
84915
Gene Name
family with sequence similarity 222 member A
Gene Aliases
C12orf34
Chromosome Location
Chr.12: 109714382 - 109770509 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK027627.1 - 1 - 2 126 129

View Full Product Details