Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDX
Entrez Gene ID
139324
Gene Name
highly divergent homeobox
Gene Aliases
CXorf43, D030011N01Rik
Chromosome Location
Chr.X: 84317874 - 84502578 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177478.1 NP_001170949.1 1 - 2 139 87
NM_001177479.1 NP_001170950.1 1 - 2 139 87
XM_011530864.1 XP_011529166.1 1 - 2 119 87
GenBank mRNA AK055240.1 - 1 - 2 113 87
BC143522.1 - 1 - 2 51 87

View Full Product Details