Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AHNAK
Entrez Gene ID
79026
Gene Name
AHNAK nucleoprotein
Gene Aliases
AHNAKRS
Chromosome Location
Chr.11: 62433542 - 62546860 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001620.2 NP_001611.1 5 - 5 13785 86
XM_005274240.1 XP_005274297.1 5 - 5 13682 86
XM_005274241.2 XP_005274298.1 5 - 5 13603 86
XM_005274245.1 XP_005274302.1 6 - 6 12371 86
XM_017018270.1 XP_016873759.1 6 - 6 13526 86
XM_017018271.1 XP_016873760.1 6 - 6 8828 86
XM_017018272.1 XP_016873761.1 6 - 6 8276 86
XM_017018273.1 XP_016873762.1 6 - 6 8276 86
GenBank mRNA AK128868.1 - 1 - 1 1508 86
X74818.1 - 1 - 1 1149 86

View Full Product Details