The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024524.3 | NP_078800.3 | 32 - 32 | 6206 | 90 |
XM_005269357.3 | XP_005269414.1 | 33 - 33 | 6317 | 90 | |
XM_011513120.2 | XP_011511422.1 | 34 - 34 | 6418 | 90 | |
XM_011513121.1 | XP_011511423.1 | 33 - 33 | 6296 | 90 | |
XM_011513122.1 | XP_011511424.1 | 33 - 33 | 5990 | 90 | |
XM_011513123.2 | XP_011511425.1 | 33 - 33 | 6333 | 90 | |
GenBank mRNA | AJ306929.1 | - | 18 - 18 | 4104 | 90 |
BC037805.1 | - | 1 - 1 | 601 | 90 | |
BC106085.1 | - | 5 - 5 | 2610 | 90 | |
Z24724.1 | - | 1 - 1 | 752 | 90 |