The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004535.2 | NP_004526.1 | 4 - 5 | 450 | 90 |
GenBank mRNA | AB020642.1 | - | 4 - 5 | 450 | 90 |
BC062313.1 | - | 4 - 5 | 304 | 90 |