The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244949.1 | NP_001231878.1 | 11 - 12 | 1324 | 69 |
NM_020918.5 | NP_065969.3 | 11 - 12 | 1320 | 69 | |
XM_005269998.1 | XP_005270055.1 | 11 - 12 | 1465 | 69 | |
GenBank mRNA | AB046780.1 | - | 5 - 6 | 612 | 69 |
AK172799.1 | - | 9 - 10 | 1153 | 69 | |
AL832464.1 | - | 11 - 12 | 1276 | 69 | |
AL833061.2 | - | 11 - 12 | 1306 | 69 | |
AL833093.2 | - | 11 - 12 | 1309 | 69 | |
BC030783.1 | - | 11 - 12 | 1288 | 69 | |
BX647451.1 | - | 11 - 12 | 1288 | 69 |