The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144653.4 | NP_653254.1 | 6 - 6 | 78 | |
XM_011518221.2 | XP_011516523.1 | 6 - 6 | 78 | ||
GenBank mRNA | AK056521.1 | - | 1 - 1 | 254 | 77 |
AK094914.1 | - | 1 - 1 | 78 |