The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001280799.1 | NP_001267728.1 | 8 - 9 | 2624 | 118 |
NM_144698.4 | NP_653299.4 | 10 - 11 | 2894 | 118 | |
GenBank mRNA | AK057853.1 | - | 1 - 2 | 784 | 118 |
AK091120.1 | - | 10 - 11 | 2859 | 118 | |
AK294308.1 | - | 8 - 9 | 2603 | 118 | |
BC101618.1 | - | 10 - 11 | 2841 | 118 | |
BC101620.1 | - | 10 - 11 | 2841 | 118 |