The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145026.3 | NP_659463.1 | 3 - 4 | 393 | 63 |
GenBank mRNA | AK058171.1 | - | 2 - 3 | 639 | 63 |
BC100956.2 | - | 2 - 3 | 448 | 63 | |
BC100957.2 | - | 2 - 3 | 448 | 63 | |
BC100958.2 | - | 2 - 3 | 448 | 63 | |
BC100959.2 | - | 3 - 4 | 542 | 63 | |
BC110065.1 | - | 2 - 3 | 448 | 63 | |
BC111414.1 | - | 2 - 3 | 448 | 63 | |
DB460008.1 | - | 3 - 4 | 394 | 63 |