Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16991655-16992159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4414842 c.2146C>A p.L716M Chr.1: 16991839-16991839 on Build GRCh38
5477800 c.2019C>T p.F673F Chr.1: 16992116-16992116 on Build GRCh38
7153961 c.2030C>T p.A677V Chr.1: 16992090-16992090 on Build GRCh38
5475575 c.2222G>T p.R741M Chr.1: 16991763-16991763 on Build GRCh38
394204 c.2228G>A p.R743H Chr.1: 16991757-16991757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772806062 Chr.1: 16992012-16992012 on Build GRCh38
rs893094160 Chr.1: 16992078-16992078 on Build GRCh38
rs371772922 Chr.1: 16991757-16991757 on Build GRCh38
rs753822981 Chr.1: 16992049-16992049 on Build GRCh38
rs989270462 Chr.1: 16991711-16991711 on Build GRCh38
rs973767624 Chr.1: 16991767-16991767 on Build GRCh38
rs751346349 Chr.1: 16991852-16991852 on Build GRCh38
rs759920062 Chr.1: 16991815-16991815 on Build GRCh38
rs17852747 Chr.1: 16991830-16991830 on Build GRCh38
rs764792843 Chr.1: 16991838-16991838 on Build GRCh38
rs774661817 Chr.1: 16991814-16991814 on Build GRCh38
rs775140060 Chr.1: 16991752-16991752 on Build GRCh38
rs143828547 Chr.1: 16992092-16992092 on Build GRCh38
rs140455242 Chr.1: 16991751-16991751 on Build GRCh38
rs749912497 Chr.1: 16991844-16991844 on Build GRCh38
rs1047502274 Chr.1: 16991678-16991678 on Build GRCh38
rs779089903 Chr.1: 16991987-16991987 on Build GRCh38
rs769856054 Chr.1: 16991719-16991719 on Build GRCh38
rs763540215 Chr.1: 16991688-16991688 on Build GRCh38
rs752442221 Chr.1: 16992037-16992037 on Build GRCh38
rs557852555 Chr.1: 16992116-16992116 on Build GRCh38
rs766723759 Chr.1: 16991730-16991730 on Build GRCh38
rs746928639 Chr.1: 16991708-16991708 on Build GRCh38
rs996052215 Chr.1: 16992056-16992056 on Build GRCh38
rs774714712 Chr.1: 16991746-16991746 on Build GRCh38
rs759858068 Chr.1: 16991979-16991980 on Build GRCh38
rs200585407 Chr.1: 16991871-16991871 on Build GRCh38
rs780063921 Chr.1: 16991703-16991703 on Build GRCh38
rs981703612 Chr.1: 16991753-16991753 on Build GRCh38
rs544545629 Chr.1: 16991834-16991834 on Build GRCh38
rs573314582 Chr.1: 16991714-16991714 on Build GRCh38
rs772483894 Chr.1: 16991995-16991995 on Build GRCh38
rs774061653 Chr.1: 16992121-16992121 on Build GRCh38
rs61999317 Chr.1: 16991825-16991825 on Build GRCh38
rs61734958 Chr.1: 16992042-16992042 on Build GRCh38
rs756039984 Chr.1: 16992040-16992040 on Build GRCh38
rs981910553 Chr.1: 16992120-16992120 on Build GRCh38
rs753237790 Chr.1: 16991750-16991750 on Build GRCh38
rs748081159 Chr.1: 16991783-16991783 on Build GRCh38
rs779034906 Chr.1: 16992059-16992059 on Build GRCh38
rs868489276 Chr.1: 16991826-16991826 on Build GRCh38
rs746079188 Chr.1: 16992061-16992061 on Build GRCh38
rs142720276 Chr.1: 16991796-16991796 on Build GRCh38
rs57809155 Chr.1: 16991953-16991953 on Build GRCh38
rs147277743 Chr.1: 16991749-16991749 on Build GRCh38
rs748290591 Chr.1: 16991715-16991715 on Build GRCh38
rs143834546 Chr.1: 16992115-16992115 on Build GRCh38
rs757300078 Chr.1: 16992052-16992052 on Build GRCh38
rs376542379 Chr.1: 16991904-16991904 on Build GRCh38
rs781206866 Chr.1: 16991772-16991772 on Build GRCh38
rs773458244 Chr.1: 16991728-16991728 on Build GRCh38
rs758564795 Chr.1: 16992063-16992063 on Build GRCh38
rs371639500 Chr.1: 16991957-16991957 on Build GRCh38
rs747361610 Chr.1: 16992083-16992083 on Build GRCh38
rs886112385 Chr.1: 16991956-16991956 on Build GRCh38
rs373300993 Chr.1: 16991692-16991692 on Build GRCh38
rs201883464 Chr.1: 16991787-16991787 on Build GRCh38
rs775779156 Chr.1: 16991996-16991996 on Build GRCh38
rs756823468 Chr.1: 16991691-16991691 on Build GRCh38
rs780888718 Chr.1: 16991902-16991902 on Build GRCh38
rs370622481 Chr.1: 16992074-16992074 on Build GRCh38
rs766036337 Chr.1: 16992017-16992017 on Build GRCh38
rs1010184962 Chr.1: 16992079-16992079 on Build GRCh38
rs754926284 Chr.1: 16991764-16991764 on Build GRCh38
rs762625030 Chr.1: 16991850-16991850 on Build GRCh38
rs937559615 Chr.1: 16991766-16991766 on Build GRCh38
rs201038868 Chr.1: 16991851-16991851 on Build GRCh38
rs774115028 Chr.1: 16992022-16992022 on Build GRCh38
rs148275181 Chr.1: 16991795-16991795 on Build GRCh38
rs1038999330 Chr.1: 16991868-16991868 on Build GRCh38
rs141058893 Chr.1: 16991702-16991702 on Build GRCh38
rs202061999 Chr.1: 16991965-16991965 on Build GRCh38
rs927414406 Chr.1: 16991805-16991805 on Build GRCh38
rs146698166 Chr.1: 16991835-16991835 on Build GRCh38
rs769228995 Chr.1: 16991747-16991747 on Build GRCh38
rs768727390 Chr.1: 16991712-16991712 on Build GRCh38
rs369509122 Chr.1: 16991976-16991976 on Build GRCh38
rs777779476 Chr.1: 16991791-16991791 on Build GRCh38
rs756062667 Chr.1: 16991964-16991964 on Build GRCh38
rs1019157778 Chr.1: 16991707-16991707 on Build GRCh38
rs769045689 Chr.1: 16992011-16992011 on Build GRCh38
rs746132666 Chr.1: 16991993-16991993 on Build GRCh38
rs762570626 Chr.1: 16992016-16992016 on Build GRCh38
rs865939676 Chr.1: 16991827-16991827 on Build GRCh38
rs913315627 Chr.1: 16991782-16991782 on Build GRCh38
rs548587977 Chr.1: 16992023-16992023 on Build GRCh38
rs930279683 Chr.1: 16991925-16991925 on Build GRCh38
rs893089570 Chr.1: 16991986-16991986 on Build GRCh38
rs769153783 Chr.1: 16992086-16992086 on Build GRCh38
rs375764016 Chr.1: 16991758-16991758 on Build GRCh38
rs971373870 Chr.1: 16992100-16992100 on Build GRCh38
rs185739088 Chr.1: 16991689-16991689 on Build GRCh38
rs528907233 Chr.1: 16991983-16991983 on Build GRCh38
rs373012377 Chr.1: 16991997-16991997 on Build GRCh38

View Full Product Details