The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301680.1 | NP_001288609.1 | 3 - 4 | 347 | 63 |
NM_001374.2 | NP_001365.1 | 3 - 4 | 370 | 63 | |
XM_011522399.1 | XP_011520701.1 | 3 - 4 | 585 | 63 | |
GenBank mRNA | AK098028.1 | - | 3 - 4 | 1207 | 63 |
AY298957.1 | - | 2 - 3 | 329 | 63 | |
BC035205.1 | - | 3 - 4 | 345 | 63 | |
BC063710.1 | - | 3 - 4 | 370 | 63 | |
U62647.1 | - | 2 - 3 | 329 | 63 |