Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS13A
Entrez Gene ID
23230
Gene Name
vacuolar protein sorting 13 homolog A
Gene Aliases
CHAC, CHOREIN
Chromosome Location
Chr.9: 77177353 - 77421537 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018037.1 NP_001018047.1 10 - 11 1019 101
NM_001018038.2 NP_001018048.1 10 - 11 1019 101
NM_015186.3 NP_056001.1 10 - 11 1019 101
NM_033305.2 NP_150648.2 10 - 11 1019 101
GenBank mRNA AB054005.1 - 10 - 11 930 101
AF337532.1 - 10 - 11 1019 101
AJ608769.1 - 10 - 11 1019 101
AJ626859.1 - 10 - 11 1019 101

View Full Product Details