The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030782.4 | NP_110409.2 | 5 - 6 | 912 | 85 |
XM_011514144.2 | XP_011512446.1 | 5 - 6 | 876 | 85 | |
GenBank mRNA | AB045223.1 | - | 5 - 6 | 669 | 85 |
AK027306.1 | - | 5 - 6 | 726 | 85 | |
BC016399.1 | - | 3 - 4 | 298 | 85 | |
BC025305.1 | - | 5 - 6 | 730 | 85 |