The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030782.4 | NP_110409.2 | 11 - 12 | 1432 | 64 |
XM_011514144.2 | XP_011512446.1 | 11 - 12 | 1396 | 64 | |
GenBank mRNA | AB045223.1 | - | 11 - 12 | 1189 | 64 |
AK027306.1 | - | 11 - 12 | 1246 | 64 | |
AK128462.1 | - | 2 - 3 | 2625 | 64 | |
BC016399.1 | - | 8 - 9 | 710 | 64 | |
BC025305.1 | - | 11 - 12 | 1250 | 64 |