Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DBN1
Entrez Gene ID
1627
Gene Name
drebrin 1
Gene Aliases
D0S117E
Chromosome Location
Chr.5: 177456612 - 177480368 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004395.3 NP_004386.2 4 - 5 499 87
NM_080881.2 NP_543157.1 5 - 6 615 87
XM_005265827.3 XP_005265884.1 4 - 5 446 87
XM_006714826.2 XP_006714889.1 4 - 5 336 87
XM_011534446.2 XP_011532748.1 5 - 6 618 87
XM_011534447.2 XP_011532749.1 4 - 5 1115 87
XM_017009139.1 XP_016864628.1 4 - 5 617 87
XM_017009140.1 XP_016864629.1 4 - 5 608 87
GenBank mRNA AK225523.1 - 4 - 5 87
AK314645.1 - 4 - 5 87
AL110225.1 - 5 - 6 87
BC000283.1 - 4 - 5 87
BC007281.1 - 4 - 5 87
BC007567.1 - 4 - 5 87
BF205663.1 - - 87
D17530.1 - 4 - 5 87
DA245346.1 - 4 - 5 87
U00802.1 - 4 - 5 87

View Full Product Details